Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASHH4
Entrez Gene ID
825166
Gene Name
histone-lysine N-methyltransferase ASHH4
Gene Aliases
AT3G59960, SDG24, SET DOMAIN PROTEIN 24, histone-lysine N-methyltransferase ASHH4
Chromosome Location
Chr.3: 22148189 - 22150583 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340008.1 NP_001319802.1 7 - 8 827 66
NM_001340009.1 NP_001326226.1 7 - 8 827 66
NM_001340010.1 NP_001326228.1 7 - 8 687 66
NM_001340011.1 NP_001326227.1 7 - 8 687 66

View Full Product Details