Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPL12
Entrez Gene ID
825173
Gene Name
squamosa promoter-binding protein-like 12
Gene Aliases
AT3G60030, squamosa promoter-binding protein-like 12
Chromosome Location
Chr.3: 22165580 - 22169984 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115866.4 NP_191562.1 9 - 10 2925 87
GenBank mRNA AB493658.1 - 9 - 10 2351 87
AJ132096.1 - 9 - 10 2351 87
BT003817.1 - 9 - 10 2351 87

View Full Product Details