Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSY5
Entrez Gene ID
825180
Gene Name
citrate synthase 5
Gene Aliases
AT3G60100, citrate synthase 5
Chromosome Location
Chr.3: 22192730 - 22196447 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340022.1 NP_001327513.1 3 - 4 308 119
NM_001340023.1 NP_001327512.1 3 - 4 308 119
NM_115873.3 NP_191569.2 3 - 4 308 119

View Full Product Details