Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIN2
Entrez Gene ID
825184
Gene Name
Glycosyl hydrolase superfamily protein
Gene Aliases
AT3G60140, BETA GLUCOSIDASE 30, BGLU30, DARK INDUCIBLE 2, SENESCENCE-RELATED GENE 2, SRG2
Chromosome Location
Chr.3: 22216540 - 22220824 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340024.1 NP_001327679.1 8 - 9 1309 69
NM_115877.4 NP_191573.1 8 - 9 1309 69

View Full Product Details