Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UCC3
Entrez Gene ID
825199
Gene Name
uclacyanin 3
Gene Aliases
AT3G60280, UCLACYANIN 3, uclacyanin 3
Chromosome Location
Chr.3: 22279485 - 22280691 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115892.4 NP_191587.1 1 - 2 213 104
GenBank mRNA AF039404.1 - 1 - 2 183 104
BT025287.1 - 1 - 2 155 104
U65650.1 - 1 - 2 194 104

View Full Product Details