Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HA7
Entrez Gene ID
825204
Gene Name
H[+]-ATPase 7
Gene Aliases
AHA7, AT3G60330, H(+)-ATPase 7, T8B10.1
Chromosome Location
Chr.3: 22298373 - 22303718 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203212.1 NP_001190141.1 11 - 12 1473 107
NM_001340039.1 NP_001327362.1 11 - 12 1466 107
NM_001340040.1 NP_001327363.1 10 - 11 1500 107
NM_115897.5 NP_191592.5 10 - 11 1471 107

View Full Product Details