Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAT3
Entrez Gene ID
825210
Gene Name
homeobox-leucine zipper protein 3
Gene Aliases
AT3G60390, HOMEOBOX PROTEIN, homeobox-leucine zipper protein 3
Chromosome Location
Chr.3: 22320580 - 22322661 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115903.3 NP_191598.1 2 - 3 846 122
GenBank mRNA AJ440781.1 - 2 - 3 555 122
AY099565.1 - 2 - 3 693 122
BT008713.1 - 2 - 3 555 122

View Full Product Details