Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABAP1
Entrez Gene ID
831145
Gene Name
ARMADILLO BTB protein 1
Gene Aliases
ARMADILLO BTB protein 1, AT5G13060, T19L5.20, T19L5_20
Chromosome Location
Chr.5: 4142910 - 4147140 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121309.6 NP_196810.5 12 - 13 1571 129
GenBank mRNA FJ422582.1 - 12 - 13 1523 129

View Full Product Details