The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_121319.5 | NP_196820.1 | 4 - 5 | 1266 | 67 |
GenBank mRNA | AF314176.1 | - | 4 - 5 | 1169 | 67 |
AF372927.1 | - | 4 - 5 | 1061 | 67 | |
AY078037.1 | - | 4 - 5 | 918 | 67 |