The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036798.1 | NP_001031875.1 | 3 - 4 | 316 | 73 |
NM_121348.4 | NP_196849.1 | 3 - 4 | 514 | 73 | |
GenBank mRNA | AK317234.1 | - | 3 - 4 | 316 | 73 |