Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FY
Entrez Gene ID
831191
Gene Name
Transducin/WD40 repeat-like superfamily protein
Gene Aliases
AT5G13480, T6I14.10, T6I14_10
Chromosome Location
Chr.5: 4326397 - 4331971 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343274.1 NP_001318555.1 11 - 12 1244 84
NM_001343275.1 NP_001331012.1 11 - 12 1175 84
NM_001343276.1 NP_001331011.1 11 - 12 1175 84
NM_121351.5 NP_196852.3 10 - 11 1211 84
GenBank mRNA BT012225.1 - 10 - 11 938 84

View Full Product Details