Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GUN5
Entrez Gene ID
831207
Gene Name
magnesium-chelatase subunit chlH, chloroplast, putative / Mg-protoporphyrin IX chelatase, putative (CHLH)
Gene Aliases
ABA-BINDING PROTEIN, ABAR, AT5G13630, CCH, CCH1, CHLH, CONDITIONAL CHLORINA, GENOMES UNCOUPLED 5, H SUBUNIT OF MG-CHELATASE, T6I14.12
Chromosome Location
Chr.5: 4387307 - 4392429 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085109.1 NP_001078578.1 3 - 4 3441 98
NM_121366.4 NP_196867.1 3 - 4 3643 98
GenBank mRNA AY070133.1 - 3 - 4 3361 98
BT002311.1 - 3 - 4 3296 98

View Full Product Details