Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZIF1
Entrez Gene ID
831219
Gene Name
zinc induced facilitator 1
Gene Aliases
AT5G13740, MSH12.21, MSH12_21, zinc induced facilitator 1
Chromosome Location
Chr.5: 4432297 - 4437148 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343291.1 NP_001331803.1 6 - 7 731 83
NM_121377.5 NP_196878.2 6 - 7 722 83

View Full Product Details