Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CARAB-AK-LYS
Entrez Gene ID
831255
Gene Name
Aspartate kinase family protein
Gene Aliases
ASPARTATE KINASE, AT5G14060, MUA22.6, MUA22_6
Chromosome Location
Chr.5: 4535367 - 4539323 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085112.2 NP_001078581.1 2 - 3 810 65
NM_001343314.1 NP_001330615.1 2 - 3 779 65
NM_121409.4 NP_196910.1 2 - 3 813 65

View Full Product Details