Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BET9
Entrez Gene ID
831277
Gene Name
bromodomain and extraterminal domain protein 9
Gene Aliases
AT5G14270, ATBET9, F18O22.60, F18O22_60, bromodomain and extraterminal domain protein 9
Chromosome Location
Chr.5: 4604608 - 4608921 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203379.1 NP_001190308.1 3 - 4 1828 77
NM_001343329.1 NP_001332743.1 4 - 5 1609 77
NM_121431.3 NP_568297.1 3 - 4 1833 77
GenBank mRNA AY059785.1 - 3 - 4 1677 77

View Full Product Details