Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYB40
Entrez Gene ID
831284
Gene Name
myb domain protein 40
Gene Aliases
AT5G14340, AtMYB40, F18O22.130, F18O22_130, myb domain protein 40
Chromosome Location
Chr.5: 4623198 - 4624960 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121438.3 NP_196938.2 2 - 3 427 85

View Full Product Details