Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CNGC18
- Entrez Gene ID
-
831339
- Gene Name
- cyclic nucleotide-gated channel 18
- Gene Aliases
-
AT5G14870, ATCNGC18, CYCLIC NUCLEOTIDE-GATED CHANNEL 18, T9L3.170, T9L3_170, cyclic nucleotide-gated channel 18
- Chromosome Location
-
Chr.5: 4808091 - 4811093 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121491.3 |
NP_196991.1 |
3 - 4 |
1184 |
126 |
View Full Product Details