Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIN8
Entrez Gene ID
831362
Gene Name
Auxin efflux carrier family protein
Gene Aliases
AT5G15100, ATPIN8, F2G14.220, F2G14_220, PIN-FORMED 8
Chromosome Location
Chr.5: 4892016 - 4894113 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121514.2 NP_197014.1 5 - 6 1207 102

View Full Product Details