Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAUT14
Entrez Gene ID
831400
Gene Name
galacturonosyltransferase 14
Gene Aliases
AT5G15470, T20K14.80, T20K14_80, galacturonosyltransferase 14
Chromosome Location
Chr.5: 5020941 - 5024750 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121551.4 NP_197051.2 3 - 4 1005 150
GenBank mRNA AK118651.1 - 3 - 4 997 150
BT005899.1 - 3 - 4 423 150
KJ138689.1 - 3 - 4 423 150

View Full Product Details