Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CXE17
Entrez Gene ID
831465
Gene Name
carboxyesterase 17
Gene Aliases
AT5G16080, AtCXE17, F1N13.220, F1N13_220, carboxyesterase 17
Chromosome Location
Chr.5: 5252251 - 5253790 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121613.3 NP_197112.1 1 - 1 1209 77
GenBank mRNA AF370327.1 - 1 - 1 1037 77
AY113907.1 - 1 - 1 986 77

View Full Product Details