Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSLA11
Entrez Gene ID
831477
Gene Name
cellulose synthase like A11
Gene Aliases
ARABIDOPSIS THALIANA CELLULOSE SYNTHASE LIKE A11, AT5G16190, ATCSLA11, T21H19.110, T21H19_110, cellulose synthase like A11
Chromosome Location
Chr.5: 5283447 - 5286842 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343440.1 NP_001330943.1 3 - 4 983 73
NM_001343441.1 NP_001330942.1 3 - 4 983 73
NM_001343442.1 NP_001330941.1 3 - 4 921 73
NM_121624.4 NP_197123.3 3 - 4 434 73
GenBank mRNA KJ138686.1 - 3 - 4 434 73

View Full Product Details