Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYN4
Entrez Gene ID
831487
Gene Name
sister chromatid cohesion 1 protein 4
Gene Aliases
ARABIDOPSIS HOMOLOG OF RAD21 3, AT5G16270, ATRAD21.3, T21H19.190, T21H19_190, sister chromatid cohesion 1 protein 4
Chromosome Location
Chr.5: 5316197 - 5322700 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121632.4 NP_197131.2 6 - 7 913 80
GenBank mRNA AF400129.1 - 6 - 7 811 80

View Full Product Details