Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TOZ
- Entrez Gene ID
-
831538
- Gene Name
- Transducin family protein / WD-40 repeat family protein
- Gene Aliases
-
AT5G16750, F5E19.90, F5E19_90, TORMOZEMBRYO DEFECTIVE
- Chromosome Location
-
Chr.5: 5504242 - 5509361 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121681.4 |
NP_568338.2 |
12 - 13 |
2228 |
154 |
View Full Product Details