Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DOT2
Entrez Gene ID
831541
Gene Name
SART-1 family
Gene Aliases
AT5G16780, DEFECTIVELY ORGANIZED TRIBUTARIES 2, DEFECTIVELY ORGANIZED TRIBUTARIES 2, F5E19.120, F5E19_120, MDF, MERISTEM-DEFECTIVE
Chromosome Location
Chr.5: 5515707 - 5522261 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343464.1 NP_001318573.1 4 - 5 1041 97
NM_001343465.1 NP_001330040.1 4 - 5 1038 97
NM_121684.4 NP_197180.1 4 - 5 1038 97

View Full Product Details