Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BPA1
Entrez Gene ID
831547
Gene Name
binding partner of acd11 1
Gene Aliases
AT5G16840, F5E19.180, F5E19_180, binding partner of acd11 1
Chromosome Location
Chr.5: 5535845 - 5538250 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085126.1 NP_001078595.1 3 - 4 360 146
NM_001085127.1 NP_001078596.1 3 - 4 296 143
NM_121690.4 NP_197186.1 3 - 4 381 143

View Full Product Details