Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARFB1B
Entrez Gene ID
831569
Gene Name
ADP-ribosylation factor B1B
Gene Aliases
ADP-ribosylation factor B1B, AT5G17060, ATARFB1B, F2K13.210, F2K13_210
Chromosome Location
Chr.5: 5610741 - 5613074 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121712.4 NP_197208.1 6 - 7 899 66
GenBank mRNA AY086691.1 - 6 - 7 834 66
AY093231.1 - 6 - 7 744 66
BT003403.1 - 6 - 7 584 66

View Full Product Details