Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDG40
Entrez Gene ID
831588
Gene Name
SET domain group 40
Gene Aliases
AT5G17240, MKP11.9, MKP11_9, SET domain group 40
Chromosome Location
Chr.5: 5666823 - 5668907 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343485.1 NP_001330251.1 1 - 2 402 142
NM_121730.4 NP_197226.2 2 - 3 323 142
GenBank mRNA AK175921.1 - 2 - 3 294 142
BT010455.1 - 2 - 3 292 142

View Full Product Details