Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BBM
Entrez Gene ID
831609
Gene Name
Integrase-type DNA-binding superfamily protein
Gene Aliases
AT5G17430, BABY BOOM, T10B6.90, T10B6_90
Chromosome Location
Chr.5: 5742393 - 5745748 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343497.1 NP_001332647.1 1 - 2 633 81
NM_121749.3 NP_197245.2 2 - 3 831 81
GenBank mRNA EF687843.1 - 2 - 3 651 81

View Full Product Details