Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APRL7
Entrez Gene ID
831930
Gene Name
APR-like 7
Gene Aliases
APR-like 7, AT5G18120, ATAPRL7, MRG7.8, MRG7_8
Chromosome Location
Chr.5: 5990909 - 5994106 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343536.1 NP_001330844.1 2 - 3 1090 77
NM_001343537.1 NP_001330845.1 1 - 2 1606 77
NM_121817.4 NP_568360.1 2 - 3 816 77
GenBank mRNA BT002852.1 - 2 - 3 627 77
BT004434.1 - 2 - 3 340 77

View Full Product Details