Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PGSIP6
Entrez Gene ID
831966
Gene Name
plant glycogenin-like starch initiation protein 6
Gene Aliases
AT5G18480, INOSITOL PHOSPHORYLCERAMIDE GLUCURONOSYLTRANSFERASE 1, IPUT1, T28N17.3, plant glycogenin-like starch initiation protein 6
Chromosome Location
Chr.5: 6131130 - 6133982 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121853.4 NP_197349.2 3 - 4 905 81
GenBank mRNA AK118951.1 - 3 - 4 746 81
KJ138680.1 - 3 - 4 710 81

View Full Product Details