Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECR1
Entrez Gene ID
832038
Gene Name
E1 C-terminal related 1
Gene Aliases
AT5G19180, E1 C-terminal related 1, T24G5.80, T24G5_80
Chromosome Location
Chr.5: 6453248 - 6456033 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343592.1 NP_001332355.1 3 - 4 607 90
NM_121923.4 NP_568370.1 3 - 4 607 90
GenBank mRNA AK226388.1 - 3 - 4 581 90
AY050426.1 - 3 - 4 544 90
AY120692.1 - 3 - 4 480 90

View Full Product Details