Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KAPP
Entrez Gene ID
832048
Gene Name
kinase associated protein phosphatase
Gene Aliases
AT5G19280, RAG1, ROOT ATTENUATED GROWTH 1, T24G5.3, kinase associated protein phosphatase
Chromosome Location
Chr.5: 6488309 - 6493439 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161248.1 NP_001154720.1 3 - 4 666 133
NM_121933.4 NP_197429.1 3 - 4 676 133
GenBank mRNA AY045853.1 - 3 - 4 622 133
AY117152.1 - 3 - 4 535 133
U09505.1 - 3 - 4 573 133

View Full Product Details