Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT24
Entrez Gene ID
832067
Gene Name
nudix hydrolase homolog 24
Gene Aliases
AT5G19470, F7K24.220, F7K24_220, NUDX24, atnudt24, nudix hydrolase homolog 24
Chromosome Location
Chr.5: 6566271 - 6568806 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343605.1 NP_001318599.1 6 - 7 847 75
NM_121952.3 NP_197448.2 6 - 7 879 75

View Full Product Details