Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSL9
Entrez Gene ID
832072
Gene Name
mechanosensitive channel of small conductance-like 9
Gene Aliases
AT5G19520, ATMSL9, T20D1.40, T20D1_40, mechanosensitive channel of small conductance-like 9
Chromosome Location
Chr.5: 6585355 - 6588966 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343609.1 NP_001331595.1 2 - 3 1697 129
NM_121957.4 NP_197453.1 2 - 3 1564 129
GenBank mRNA AK228195.1 - 2 - 3 1498 129
BT006457.1 - 1 - 2 1362 129

View Full Product Details