Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ROPGEF10
Entrez Gene ID
832076
Gene Name
ROP uanine nucleotide exchange factor 10
Gene Aliases
ARABIDOPSIS THALIANA ROP UANINE NUCLEOTIDE EXCHANGE FACTOR 10, AT5G19560, ATROPGEF10, GEF10, ROP (rho of plants) guanine nucleotide exchange factor 10, ROP uanine nucleotide exchange factor 10, T20D1.80, T20D1_80, guanine exchange factor 10
Chromosome Location
Chr.5: 6602910 - 6606506 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343611.1 NP_001318601.1 3 - 4 430 72
NM_001343612.1 NP_001331396.1 2 - 3 625 72
NM_001343613.1 NP_001331395.1 2 - 3 625 72
NM_001343614.1 NP_001331397.1 2 - 3 625 72
NM_121961.2 NP_197457.1 2 - 3 244 72
GenBank mRNA DQ459196.1 - 2 - 3 244 72

View Full Product Details