Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COQ3
Entrez Gene ID
817642
Gene Name
coenzyme Q 3
Gene Aliases
AT2G30920, ATCOQ3, EMB3002, F7F1.13, F7F1_13, coenzyme Q 3, embryo defective 3002
Chromosome Location
Chr.2: 13157089 - 13160886 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336307.1 NP_001324991.1 3 - 4 630 94
NM_128644.4 NP_180649.1 2 - 3 480 94
GenBank mRNA AY045828.1 - 2 - 3 471 94
AY091366.1 - 2 - 3 363 94

View Full Product Details