Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SKdZeta
Entrez Gene ID
817649
Gene Name
SHAGGY-related protein kinase dZeta
Gene Aliases
A. THALIANA SHAGGY-LIKE KINASE GROUP 2 2, ASKdZeta, AT2G30980, ATSK2-2, ATSK23, BIL1, BIN2-LIKE 1, F7F1.19, F7F1_19, SHAGGY-LIKE PROTEIN KINASE 23, SHAGGY-related protein kinase dZeta
Chromosome Location
Chr.2: 13182025 - 13185973 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128652.4 NP_180655.1 2 - 3 301 95
GenBank mRNA AY064020.1 - 2 - 3 251 95
AY094423.1 - 2 - 3 277 95
AY096698.1 - 2 - 3 198 95

View Full Product Details