Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CDT1A
- Entrez Gene ID
-
817684
- Gene Name
- CDT1-like protein A
- Gene Aliases
-
ARABIDOPSIS HOMOLOG OF YEAST CDT1, ARABIDOPSIS HOMOLOG OF YEAST CDT1 A, AT2G31270, ATCDT1A, CDT1, F16D14.11, F16D14_11, homolog of yeast CDT1 A
- Chromosome Location
-
Chr.2: 13328862 - 13331831 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128683.4 |
NP_180685.1 |
5 - 6 |
1710 |
89 |
View Full Product Details