Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UGT74D1
Entrez Gene ID
817732
Gene Name
UDP-glucosyl transferase 74D1
Gene Aliases
AT2G31750, F20M17.21, F20M17_21, UDP-glucosyl transferase 74D1
Chromosome Location
Chr.2: 13497118 - 13500228 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336347.1 NP_001325305.1 1 - 2 727 89
NM_128733.5 NP_180734.1 1 - 2 842 89
GenBank mRNA AY125506.1 - 1 - 2 683 89
BT000622.1 - 1 - 2 648 89
DQ158907.1 - 1 - 2 648 89
KJ138885.1 - 1 - 2 648 89

View Full Product Details