Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLP23
Entrez Gene ID
817828
Gene Name
receptor like protein 23
Gene Aliases
AT2G32680, AtRLP23, F24L7.18, F24L7_18, receptor like protein 23
Chromosome Location
Chr.2: 13859769 - 13862614 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128827.2 NP_180827.1 1 - 1 1638 63

View Full Product Details