Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BGAL9
Entrez Gene ID
817842
Gene Name
beta galactosidase 9
Gene Aliases
AT2G32810, F24L7.5, F24L7_5, beta galactosidase 9, beta-galactosidase 9
Chromosome Location
Chr.2: 13919127 - 13925421 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202731.1 NP_001189660.1 6 - 7 740 97
NM_128841.5 NP_565755.1 6 - 7 810 97
GenBank mRNA AJ270305.1 - 6 - 7 714 97

View Full Product Details