Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFN2
Entrez Gene ID
817855
Gene Name
zinc finger nuclease 2
Gene Aliases
AT2G32930, T21L14.13, T21L14_13, zinc finger nuclease 2
Chromosome Location
Chr.2: 13965714 - 13968957 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336416.1 NP_001318339.1 4 - 5 938 89
NM_001336417.1 NP_001325104.1 3 - 4 433 89
NM_128853.2 NP_565758.1 4 - 5 926 89
GenBank mRNA AF138744.1 - 4 - 5 388 89

View Full Product Details