Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XID
Entrez Gene ID
817886
Gene Name
myosin XI D
Gene Aliases
AT2G33240, ATXID, F25I18.2, F25I18_2, MYOSIN XI D, myosin XI D
Chromosome Location
Chr.2: 14086584 - 14097285 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336434.1 NP_001324212.1 38 - 39 5397 127
NM_001336435.1 NP_001324208.1 38 - 39 4943 127
NM_001336436.1 NP_001324211.1 38 - 39 4964 127
NM_001336437.1 NP_001324213.1 24 - 25 3454 127
NM_001336438.1 NP_001324210.1 38 - 39 5225 127
NM_001336439.1 NP_001324209.1 39 - 40 4919 127
NM_001336440.1 NP_001324214.1 39 - 40 4919 127
NM_128883.3 NP_180882.2 38 - 39 4982 127
GenBank mRNA LC177513.1 - 37 - 38 4910 127

View Full Product Details