Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CPL3
- Entrez Gene ID
-
817919
- Gene Name
- C-terminal domain phosphatase-like 3
- Gene Aliases
-
AT2G33540, ATCPL3, C-terminal domain phosphatase-like 3, F4P9.31, F4P9_31
- Chromosome Location
-
Chr.2: 14203401 - 14209192 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128914.3 |
NP_180912.2 |
1 - 2 |
1013 |
93 |
View Full Product Details