The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202741.2 | NP_001189670.2 | 4 - 5 | 647 | 111 |
NM_001336484.1 | NP_001323468.1 | 4 - 5 | 632 | 111 | |
NM_001336485.1 | NP_001323467.1 | 4 - 5 | 527 | 111 | |
NM_128960.3 | NP_180956.2 | 4 - 5 | 622 | 111 | |
NM_201866.2 | NP_973595.1 | 4 - 5 | 622 | 111 | |
GenBank mRNA | BT002072.1 | - | 4 - 5 | 551 | 111 |
BT015789.1 | - | 4 - 5 | 527 | 111 |