Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEE18
Entrez Gene ID
817970
Gene Name
maternal effect embryo arrest 18
Gene Aliases
AT2G34090, T14G11.21, T14G11_21, maternal effect embryo arrest 18
Chromosome Location
Chr.2: 14396016 - 14399275 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202741.2 NP_001189670.2 9 - 10 1098 82
NM_001336484.1 NP_001323468.1 9 - 10 1083 82
NM_001336485.1 NP_001323467.1 9 - 10 978 82
NM_128960.3 NP_180956.2 9 - 10 1073 82
NM_201866.2 NP_973595.1 9 - 10 1073 82
GenBank mRNA BT002072.1 - 9 - 10 1002 82
BT015789.1 - 9 - 10 978 82

View Full Product Details