Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP6
Entrez Gene ID
841596
Gene Name
ubiquitin-specific protease 6
Gene Aliases
AT1G51710, ATUBP6, F19C24.8, F19C24_8, UBIQUITIN-SPECIFIC PROTEASE 6, ubiquitin-specific protease 6
Chromosome Location
Chr.1: 19175609 - 19180195 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084229.1 NP_001077698.1 5 - 6 473 90
NM_104049.4 NP_564596.1 7 - 8 887 90
GenBank mRNA AY050817.1 - 7 - 8 651 90
AY084730.1 - 7 - 8 655 90
AY114081.1 - 6 - 7 586 90
BT000658.1 - 7 - 8 656 90

View Full Product Details