Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYP81
Entrez Gene ID
841599
Gene Name
syntaxin of plants 81
Gene Aliases
ARABIDOPSIS THALIANA ORTHOLOG OF YEAST UFE1 (UNKNOWN FUNCTION-ESSENTIAL 1), AT1G51740, ATSYP81, ATUFE1, F19C24.5, F19C24_5, ORTHOLOG OF YEAST UFE1 (UNKNOWN FUNCTION-ESSENTIAL 1), UFE1, syntaxin of plants 81
Chromosome Location
Chr.1: 19188799 - 19191372 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333499.1 NP_001323219.1 1 - 2 348 90
NM_104052.3 NP_564597.1 1 - 2 304 90
GenBank mRNA BT002917.1 - 1 - 2 273 90
BT004352.1 - 1 - 2 84 90

View Full Product Details