Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GDAP1
Entrez Gene ID
825330
Gene Name
GRIP-related ARF-binding domain-containing protein 1
Gene Aliases
AT3G61570, GC3, GOLGIN CANDIDATE 3, GRIP-related ARF-binding domain-containing protein 1
Chromosome Location
Chr.3: 22779441 - 22783938 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340110.1 NP_001327121.1 3 - 4 642 80
NM_116022.5 NP_191716.2 3 - 4 719 80
GenBank mRNA AK226722.1 - 3 - 4 582 80
BT002756.1 - 3 - 4 423 80
EU249332.1 - 3 - 4 423 80

View Full Product Details