Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG6
Entrez Gene ID
825344
Gene Name
AUTOPHAGY 6
Gene Aliases
AT3G61710, ATATG6, AUTOPHAGY 6, AtBECLIN1, BECLIN1, F21F14.6
Chromosome Location
Chr.3: 22839232 - 22842446 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035837.1 NP_001030914.1 3 - 4 839 142
NM_001340123.1 NP_001327722.1 3 - 4 861 142
NM_116036.4 NP_567116.1 3 - 4 861 142
NM_202746.2 NP_974475.1 3 - 4 847 142
GenBank mRNA AY039613.1 - 3 - 4 847 142
BT000508.1 - 3 - 4 668 142

View Full Product Details